Nalpha-Boc-Nepsilon-Fmoc-L-lysine, 98%
Nalpha-Boc-Nepsilon-Fmoc-L-lysine, 98%
Nalpha-Boc-Nepsilon-Fmoc-L-lysine, 98%
Thermo Scientific Chemicals

Nalpha-Boc-Nepsilon-Fmoc-L-lysine, 98%

CAS: 84624-27-1 | C26H32N2O6 | 468.55 g/mol
Quantity:
5 g
25 g
Catalog number H61126.06
also known as H61126-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS84624-27-1
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular FormulaC26H32N2O6
InChI KeyJYEVQYFWINBXJU-ANBDAQEENA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.

Solubility
Soluble in dimethylformamide. Insoluble in water.

Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jong Eun Oh.; Keun Hyeung Lee. Synthesis of novel unnatural amino acid as a building block and its incorporation into an antimicrobial peptide. Bioorganic & Medicinal Chemistry. 1999, 7 (12), 2985-2990.
  2. Steve R. Bull.; Mustafa O. Guler.; Rafael E. Bras.; Thomas J. Meade.; Samuel I. Stupp. Self-Assembled Peptide Amphiphile Nanofibers Conjugated to MRI Contrast Agents. Nano Lett. 2005, 5 (1), 1-4.