6-Heptyn-1-ol, 95%
6-Heptyn-1-ol, 95%
6-Heptyn-1-ol, 95%
Thermo Scientific Chemicals

6-Heptyn-1-ol, 95%

CAS: 63478-76-2 | C7H12O | 112.17 g/mol
Quantity:
1 g
5 g
Catalog number H61509.03
also known as H61509-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS63478-76-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>94.0%
6-Heptyn-1-ol is used as a reagent in in the preparation of many cyclic compounds. It is also used as a Intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Heptyn-1-ol is used as a reagent in in the preparation of many cyclic compounds. It is also used as a Intermediate in organic synthesis.

Solubility
Soluble in chloroform, dichloromethane and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Shengming Ma; Ei-ichi Negishi. Anti-Carbometalation of Homopropargyl Alcohols and Their Higher Homologues via Non-Chelation-Controlled Syn-Carbometalation and Chelation-Controlled Isomerization. J. Org. Chem. 1997, 62(4), 784-785.
  2. H. Menzel; S. Horstmann; M.D. Mowery; M. Caic; C.E. Evans. Diacetylene polymerization in self-assembled monolayers: influence of the odd/even nature of the methylene spacer. Polymer. 2000, 41(22), 8113-8119.