N-Boc-3-(4-pyridyl)-D-alanine, 95%
N-Boc-3-(4-pyridyl)-D-alanine, 95%
N-Boc-3-(4-pyridyl)-D-alanine, 95%
Thermo Scientific Chemicals

N-Boc-3-(4-pyridyl)-D-alanine, 95%

CAS: 37535-58-3 | C13H18N2O4 | 266.297 g/mol
Quantity:
1 g
5 g
Catalog number H62215.03
also known as H62215-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS37535-58-3
IUPAC Name(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoate
Molecular FormulaC13H17N2O4
InChI KeyFNYWDMKESUACOU-SNVBAGLBSA-M
SMILESCC(C)(C)OC(=O)N[C@H](CC1=CC=NC=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only