cis-4-(Boc-amino)cyclohexanol, 97%
cis-4-(Boc-amino)cyclohexanol, 97%
cis-4-(Boc-amino)cyclohexanol, 97%
cis-4-(Boc-amino)cyclohexanol, 97%
Thermo Scientific Chemicals

cis-4-(Boc-amino)cyclohexanol, 97%

CAS: 167081-25-6 | C11H21NO3 | 215.29 g/mol
Quantity:
250 mg
1 g
5 g
Catalog number H62238.06
also known as H62238-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Specifications
CAS167081-25-6
Chemical Name or Materialcis-4-(Boc-amino)cyclohexanol
Formula Weight215.28
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
cis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents

Solubility
Slightly soluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only