6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine, 96%, Thermo Scientific Chemicals
6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine, 96%, Thermo Scientific Chemicals
6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine, 96%, Thermo Scientific Chemicals

Catalog NumberQuantity
H62259.03
also known as H62259-03
1 g
Catalog number H62259.03
also known as H62259-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS1160102-28-2
Chemical Name or Material6-Bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine
Formula Weight232.05
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only