N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%
N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%
N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%
N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%
Thermo Scientific Chemicals

N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%

CAS: 134098-70-7 | C23H25NO6 | 411.454 g/mol
Quantity:
1 g
5 g
Catalog number H62680.03
also known as H62680-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS134098-70-7
IUPAC Name4-(tert-butoxy)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
Molecular FormulaC23H25NO6
InChI KeyVZXQYACYLGRQJU-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
N-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only