3,5-Dichlorophenylacetic acid, 95%
3,5-Dichlorophenylacetic acid, 95%
3,5-Dichlorophenylacetic acid, 95%
Thermo Scientific Chemicals

3,5-Dichlorophenylacetic acid, 95%

CAS: 51719-65-4 | C8H6Cl2O2 | 205.034 g/mol
Quantity:
1 g
5 g
Catalog number H63293.03
also known as H63293-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS51719-65-4
IUPAC Name2-(3,5-dichlorophenyl)acetic acid
Molecular FormulaC8H6Cl2O2
InChI KeyRERINLRFXYGZEE-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC(Cl)=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only