Methyl 2,5-difluoro-4-nitrobenzoate, 95%
Methyl 2,5-difluoro-4-nitrobenzoate, 95%
Methyl 2,5-difluoro-4-nitrobenzoate, 95%
Thermo Scientific Chemicals

Methyl 2,5-difluoro-4-nitrobenzoate, 95%

CAS: 924868-81-5 | C8H5F2NO4 | 217.13 g/mol
Quantity:
1 g
5 g
Catalog number H63472.06
also known as H63472-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS924868-81-5
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only