4-Chloro-L-phenylalanine, 97%
4-Chloro-L-phenylalanine, 97%
4-Chloro-L-phenylalanine, 97%
Thermo Scientific Chemicals

4-Chloro-L-phenylalanine, 97%

CAS: 14173-39-8 | C9H10ClNO2 | 199.63 g/mol
Quantity:
250 mg
1 g
5 g
Catalog number H63573.03
also known as H63573-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS14173-39-8
IUPAC Name2-amino-3-(4-chlorophenyl)propanoic acid
Molecular FormulaC9H10ClNO2
InChI KeyNIGWMJHCCYYCSF-UHFFFAOYNA-N
SMILESNC(CC1=CC=C(Cl)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
4-Chloro-L-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-L-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in acetic acid. Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. T Yoshino; I Yamaguchi. Possible involvement of 5-HT2 receptor activation in aggravation of diet-induced acute pancreatitis in mice. Journal of Pharmacology and Experimental Therapeutics. 1997, 283 (3), 1495-1502.
  2. O Benkert, M Bender, F Bidlingmaier, O Butenandt. Effect of p-chlorophenylalanine (PCPA) on pituitary hormones and testosterone in the human. Arzneimittel-Forschung. 1976, 26 (7), 1369-1371 .