(R)-2-(Boc-amino)butyric acid, 95%
(R)-2-(Boc-amino)butyric acid, 95%
(R)-2-(Boc-amino)butyric acid, 95%
Thermo Scientific Chemicals

(R)-2-(Boc-amino)butyric acid, 95%

CAS: 45121-22-0 | C9H17NO4 | 203.238 g/mol
Quantity:
1 g
5 g
25 g
Catalog number H63991.06
also known as H63991-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS45121-22-0
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Molecular FormulaC9H17NO4
InChI KeyPNFVIPIQXAIUAY-UHFFFAOYNA-N
SMILESCCC(NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.

Solubility
Soluble in DMF (1mmol in 1ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

General References:

  1. Yoo Lim Kam,; Hee-Kyung Rhee,; Hyewhon Rhim,; Seung Keun Back,; Heung Sik Na,; Hea-Young Park Choo. Synthesis and T-type calcium channel blocking activity of novel diphenylpiperazine compounds, and evaluation of in vivo analgesic activity. Bioorganic & Medicinal Chemistry. 2010, 18 (16), 5938-5944.