5-Methylpyridine-2-carbonitrile, 97+%
5-Methylpyridine-2-carbonitrile, 97+%
5-Methylpyridine-2-carbonitrile, 97+%
Thermo Scientific Chemicals

5-Methylpyridine-2-carbonitrile, 97+%

CAS: 1620-77-5 | C7H6N2 | 118.14 g/mol
Quantity:
5 g
25 g
Catalog number H64539.06
also known as H64539-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1620-77-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
FormSolid
Assay from Suppliers CofA≥97.0% (GC)
Melting Point69-77°C
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only