Thermo Scientific Chemicals

Methoxyacetic anhydride, 95%, Thermo Scientific Chemicals

Catalog number: H64541.14
25 g, Each
Thermo Scientific Chemicals

Methoxyacetic anhydride, 95%, Thermo Scientific Chemicals

Catalog number: H64541.14
25 g, Each
Quantity
Catalog number: H64541.14
also known as H64541-14
Price (USD)
Price: 98.20
Online price: 84.65
Your price:
Quantity
-

Chemical Identifiers

CAS
19500-95-9
IUPAC Name
2-methoxyacetyl 2-methoxyacetate
Molecular Formula
C6H10O5
InChI Key
PEHFQQWAINXOQG-UHFFFAOYSA-N
SMILES
COCC(=O)OC(=O)COC
Assay (GC)
>94.0%

Description

Methoxyacetic anhydride is used as the intermediate of 5-Aza-2'-deoxycytidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetic anhydride is used as the intermediate of 5-Aza-2′-deoxycytidine.

Solubility
Very Soluble in water.(193 g/L) (25°C)

Notes
Moisture sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text