L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride, 98%
L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride, 98%
L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride, 98%
L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride, 98%

CAS: 94347-11-2 | C15H22ClNO4 | 315.79 g/mol
Quantity:
1 g
5 g
Catalog number H65937.06
also known as H65937-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Specifications
Beilstein2538326
CAS94347-11-2
Chemical Name or MaterialL-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride
ColorWhite
Formula Weight315.79
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only