Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine, 95%
Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine, 95%
Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine, 95%
Thermo Scientific Chemicals

Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine, 95%

CAS: 201485-17-8 | C22H25N3O5 | 411.46 g/mol
Quantity:
1 g
5 g
Catalog number H66189.03
also known as H66189-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS201485-17-8
IUPAC Name(2S)-6-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular FormulaC22H25N3O5
InChI KeyVJZUCXPETVPQIB-TWYLJJHKNA-N
SMILESNC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only