3-[2-(Ethoxycarbonyl)ethyl]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-[2-(Ethoxycarbonyl)ethyl]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Quantity:
1 g
5 g
Catalog number H66350.03
also known as H66350-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein20464729
CAS935854-36-7
Chemical Name or Material3-[2-(Ethoxycarbonyl)ethyl]benzeneboronic acid pinacol ester
ColorWhite to Yellow
Formula Weight304.19
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only