5-Bromopyrazine-2-carboxylic acid, 95%
5-Bromopyrazine-2-carboxylic acid, 95%
5-Bromopyrazine-2-carboxylic acid, 95%
Thermo Scientific Chemicals

5-Bromopyrazine-2-carboxylic acid, 95%

CAS: 876161-05-6 | C5H3BrN2O2 | 203.00 g/mol
Quantity:
250 mg
1 g
Catalog number H66382.03
also known as H66382-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS876161-05-6
IUPAC Name5-bromopyrazine-2-carboxylic acid
Molecular FormulaC5H3BrN2O2
InChI KeyNPJHCKULJAIWGV-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=C(Br)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)95.40%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only