6-Chloro-7-azaindole, 95%
6-Chloro-7-azaindole, 95%
6-Chloro-7-azaindole, 95%
6-Chloro-7-azaindole, 95%
Thermo Scientific Chemicals

6-Chloro-7-azaindole, 95%

CAS: 55052-27-2 | C7H5ClN2 | 152.58 g/mol
Quantity:
1 g
5 g
Catalog number H66655.03
also known as H66655-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS55052-27-2
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥94.0%
Appearance (Color)Pale cream
Melting Point170-175?C
FormFused solid
Proton NMRConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only