4-Amino-6-methylpyrimidine, 95%
4-Amino-6-methylpyrimidine, 95%
4-Amino-6-methylpyrimidine, 95%
Thermo Scientific Chemicals

4-Amino-6-methylpyrimidine, 95%

CAS: 3435-28-7 | C5H7N3 | 109.132 g/mol
Catalog NumberQuantity
H66832.MD
also known as H66832-MD
250 mg
Catalog number H66832.MD
also known as H66832-MD
Price (JPY)
-
Quantity:
250 mg
Request bulk or custom format
Specifications
Beilstein116647
CAS3435-28-7
Chemical Name or Material4-Amino-6-methylpyrimidine
Formula Weight109.13
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only