PBDB-T-2Cl, Thermo Scientific Chemicals
PBDB-T-2Cl, Thermo Scientific Chemicals
Thermo Scientific Chemicals

PBDB-T-2Cl, Thermo Scientific Chemicals

Quantity:
100 mg
250 mg
500 mg
Catalog number H66867.ME
also known as H66867-ME
Price (JPY)
-
Request A Quote
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS2239295-71-5
SpecificationsSpecification SheetSpecification Sheet
Bandgap energyt.b.d.
Appearance (Color)Black
CommentWhen bandgap is measured it will be added.
FormFlake or threadlike
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only