Iodine Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Iodine Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Iodine Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Iodine Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

I2, CAS Number-7553-56-2, tincture iodine, iodine sublimed, iodine solution, iodine crystals, iodine, iode, diiodine, vistarin, iodio, eranol, 1L, CHEBI:17606, Brown, PNDPGZBMCMUPRI-UHFFFAOYSA-N, molecular iodine, 0 deg.C, 11355, 253.809, Amber glass bottle, Liquid, 807, II, 0.7, 14mmHg
Catalog NumberQuantity
J-4410-15
also known as J/4410/15
1 L
Catalog number J-4410-15
also known as J/4410/15
Price (JPY)
-
Quantity:
1 L
Request bulk or custom format
Specifications
CAS7553-56-2
ChEBICHEBI:17606
ColorBrown
IUPAC Namemolecular iodine
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
View more
I2, CAS Number-7553-56-2, tincture iodine, iodine sublimed, iodine solution, iodine crystals, iodine, iode, diiodine, vistarin, iodio, eranol, 1L, CHEBI:17606, Brown, PNDPGZBMCMUPRI-UHFFFAOYSA-N, molecular iodine, 0 deg.C, 11355, 253.809, Amber glass bottle, Liquid, 807, II, 0.7, 14mmHg