NP-40 lysis buffer
Thermo Scientific Chemicals

NP-40 lysis buffer

Quantity:
250 mL
500 mL
Catalog number J60766.AP
also known as J60766-AP
Price (JPY)
-
Request A Quote
Quantity:
500 mL
Request bulk or custom format
Specifications
ColorColorless
FormatLiquid
Quantity500 mL
Unit SizeEach
NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.

Solubility
It is soluble in water.

Notes
This buffer contains: 50mM Tris-HCl (pH 7.4), 150mM NaCl, 1%NP-40 and 5mM EDTA. To use, incubate cells in 5 volumes of buffer on ice for 10-30 minutes.
RUO – Research Use Only

General References:

  1. M Herrmann.; HM Lorenz.; R Voll.; M Grünke. A rapid and simple method for the isolation of apoptotic DNA fragments. Nucleic Acids Res. 1994, 22,(24), 5506-5507.
  2. K Guo.; J Wang.; V Andres.; RC Smith. MyoD-induced expression of p21 inhibits cyclin-dependent kinase activity upon myocyte terminal differentiation. Mol. Cell. Biol. 1995, 15,(7), 3823-3829.
Contents & Storage
Keep cold