(-)-Epicatechin gallate, Thermo Scientific Chemicals
(-)-Epicatechin gallate, Thermo Scientific Chemicals
(-)-Epicatechin gallate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(-)-Epicatechin gallate, Thermo Scientific Chemicals

A natural product from green tea that induces apoptosis
Quantity:
10 mg
20 mg
Catalog number J60814.FPL
also known as J60814-F+
Price (JPY)
-
Quantity:
20 mg
Request bulk or custom format
Chemical Identifiers
CAS1257-08-5
IUPAC Name(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-1-benzopyran-3-yl 3,4,5-trihydroxybenzoate
Molecular FormulaC22H18O10
InChI KeyLSHVYAFMTMFKBA-TZIWHRDSSA-N
SMILESOC1=CC(O)=C2C[C@@H](OC(=O)C3=CC(O)=C(O)C(O)=C3)[C@H](OC2=C1)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder
Assay from Supplier's CofA≥97.0% (HPLC)
(-)-Epicatechin gallate is used as an antioxidant investigated as an adjunct treatment of some methyicillin resistant bacteria. Epicatechin gallate binds to the cell membrane in methicillin-resistant Staphylococcus aureus which decreases the fluidity of the bilayer and induces changes in gene expression. One of the catechin isomers and a potent antioxidant that can modulate a wide range of membrane proteins. Its bilayer-modifying potency was tested using gramicidin A (gA) channels as probes. All the catechins alter gA channel function and modify bilayer properties, with a 500-fold range in potency.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Epicatechin gallate is used as an antioxidant investigated as an adjunct treatment of some methyicillin resistant bacteria. Epicatechin gallate binds to the cell membrane in methicillin-resistant Staphylococcus aureus which decreases the fluidity of the bilayer and induces changes in gene expression. One of the catechin isomers and a potent antioxidant that can modulate a wide range of membrane proteins. Its bilayer-modifying potency was tested using gramicidin A (gA) channels as probes. All the catechins alter gA channel function and modify bilayer properties, with a 500-fold range in potency.

Solubility
Soluble in water, acetone, DMSO, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
RUO – Research Use Only

General References:

  1. SJ Baek.; JS Kim.; FR Jackson.; TE Eling. Epicatechin gallate-induced expression of NAG-1 is associated with growth inhibition and apoptosis in colon cancer cells. Carcinogenesis . 2015, 25,(12), S254-S296.
  2. CT Ho.; Q Chen.; H Shi.; KQ Zhang.; RT Rosen. Antioxidative effect of polyphenol extract prepared from various Chinese teas. Preventive medicine . 1992, 21,(4), 520-525.