Potassium acetate, 1M aq. soln., pH 7.5, RNAse free, Thermo Scientific Chemicals
Potassium acetate, 1M aq. soln., pH 7.5, RNAse free, Thermo Scientific Chemicals
Potassium acetate, 1M aq. soln., pH 7.5, RNAse free, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium acetate, 1M aq. soln., pH 7.5, RNAse free, Thermo Scientific Chemicals

Catalog NumberQuantity
J60832.AK
also known as J60832-AK
250 mL
Catalog number J60832.AK
also known as J60832-AK
Price (JPY)
-
Quantity:
250 mL
Request bulk or custom format
Specifications
CAS127-08-2
ChEBICHEBI:32029
Chemical Name or MaterialPotassium acetate
ColorColorless
Concentration or Composition (by Analyte or Components)1M aq. soln.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only