Nuclear Fast Red Soln., enhanced stability
Thermo Scientific™

Nuclear Fast Red Soln., enhanced stability

Quantity:
500 mL
1000 mL
Catalog number J61010.AU
also known as J61010-AU
Price (JPY)
-
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialNuclear Fast Red Soln.
Health Hazard 1H318
Health Hazard 3P280-P305+P351+P338-P310
PackagingPlastic bottle
Physical FormLiquid
View more
A histological counterstain. Nuclear Fast Red solution turns cell nuclei red and cytoplasm a pale pink.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only