Agarose, high melting temperature, medium resolution
Agarose, high melting temperature, medium resolution
Agarose, high melting temperature, medium resolution
Thermo Scientific Chemicals

Agarose, high melting temperature, medium resolution

For standard DNA applications | CAS: 9012-36-6
Quantity:
25 g
100 g
500 g
Catalog number J61123.14
also known as J61123-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS9012-36-6
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H38O19
InChI KeyMJQHZNBUODTQTK-WKGBVCLCSA-N
SMILESOC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Gel Strength 1.0% >1200 g/cm²: Conforms
FormPowder
Appearance (Color)White
Assay (unspecified)Gelling Range 36-39 °C: Conforms
For standard DNA applications.Agarose, High Melting Temperature, Medium Resolution is used for standard DNA separations. It is involved in the purification of protein by using column-based preparative scale separation as in gel filtration chromatography and ion exchange chromatography

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For standard DNA applications.Agarose, High Melting Temperature, Medium Resolution is used for standard DNA separations. It is involved in the purification of protein by using column-based preparative scale separation as in gel filtration chromatography and ion exchange chromatography

Solubility
Soluble in water.

Notes
Melting temp: ∼88°.Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Hernández, C.; Alvarez, C.; González, C.; Ayala, M. S.; León, C. M.; Ramírez, J. D. Identification of Six New World Leishmania species through the implementation of a High-Resolution Melting (HRM) genotyping assay. Parasites Vectors 2014, 7 (1), 1-7.
  2. Alnuaimi, A. D.; Wiesenfeld, D.; O'Brien-Simpson, N. M.; Reynolds, E. C.; Peng, B.; McCullough, M. J. The development and validation of a rapid genetic method for species identification and genotyping of medically important fungal pathogens using high-resolution melting curve analysis. Mol Oral Microbiol 2014, 29 (3), 117-130.