Nicardipine hydrochloride, 98%
Nicardipine hydrochloride, 98%
Nicardipine hydrochloride, 98%
Thermo Scientific Chemicals

Nicardipine hydrochloride, 98%

L-type calcium channel blocker | CAS: 54527-84-3 | C26H30ClN3O6 | 515.991 g/mol
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61269.03
also known as J61269-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS54527-84-3
IUPAC Namehydrogen 3-{2-[benzyl(methyl)amino]ethyl} 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate chloride
Molecular FormulaC26H30ClN3O6
InChI KeyAIKVCUNQWYTVTO-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OCCN(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Suppliers CofA≥97.5%
L-type calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only