Thermo Scientific Chemicals
Furosemide, 97+%, Thermo Scientific Chemicals
Catalog number: J61457.09
10 g, Each
Thermo Scientific Chemicals
Furosemide, 97+%, Thermo Scientific Chemicals
Catalog number: J61457.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61457.09
also known as J61457-09
Price (USD)
Price: 164.00
Online price: 139.65
Your price:
Quantity
-
Description
Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter