Thermo Scientific Chemicals

MES-buffered saline (5X), pH 6.5

Catalog number: J61979.AP
500 mL, Each
Thermo Scientific Chemicals

MES-buffered saline (5X), pH 6.5

Catalog number: J61979.AP
500 mL, Each
Quantity
Catalog number: J61979.AP
also known as J61979-AP
Price (USD)
Price: 160.00
Online price: 136.65
Your price:
Quantity
-

Chemical Identifiers

CAS
145224-94-8
IUPAC Name
4-(2-sulfonatoethyl)morpholin-4-ium
Molecular Formula
C6H13NO4S
InChI Key
SXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES
[O-]S(=O)(=O)CC[NH+]1CCOCC1
Appearance (Color)
Clear colorless
Composition
Sodium Chloride: 750 mM
Composition
MES: 125 mM
pH
6.3-6.7
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text