Triolein, tech.
Triolein, tech.
Triolein, tech.
Thermo Scientific Chemicals

Triolein, tech.

CAS: 122-32-7 | C57H104O6 | 885.453 g/mol
Quantity:
25 g
100 g
Catalog number J62419.14
also known as J62419-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS122-32-7
IUPAC Name1,3-bis[(9Z)-octadec-9-enoyloxy]propan-2-yl (9Z)-octadec-9-enoate
Molecular FormulaC57H104O6
InChI KeyPHYFQTYBJUILEZ-IUPFWZBJSA-N
SMILESCCCCCCCC\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
Identification (FTIR)Conforms
FormLiquid
Proton NMRConforms
Glycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.

Solubility
Soluble in chlorofom, ether, and carbon tetrachloride

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C
RUO – Research Use Only

General References:

  1. A Salis.; M Pinna.; M Monduzzi.; V Solinas. Biodiesel production from triolein and short chain alcohols through biocatalysis. Journal of Biotechnology. 2005, 119, (3), 291-299.
  2. F Mori.; JC Lim.; OG Raney.; CM Elsik.; CA Miller. Phase behavior, dynamic contacting and detergency in systems containing triolein and nonionic surfactants. Colloids and surfaces. 1989, 40, (1), 323-345.