CAPS, 0.5M buffer soln., pH 10.5
CAPS, 0.5M buffer soln., pH 10.5
CAPS, 0.5M buffer soln., pH 10.5
CAPS, 0.5M buffer soln., pH 10.5
Thermo Scientific Chemicals

CAPS, 0.5M buffer soln., pH 10.5

CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol
Quantity:
250 mL
500 mL
Catalog number J62446.AK
also known as J62446-AK
Price (JPY)
-
Request A Quote
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS1135-40-6
IUPAC Name3-(cyclohexylamino)propane-1-sulfonic acid
Molecular FormulaC9H19NO3S
InChI KeyPJWWRFATQTVXHA-UHFFFAOYSA-N
SMILESOS(=O)(=O)CCCNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
CompositionCAPS: 0.5M
Appearance (Color)Clear colorless
FormLiquid
pH10.3-10.7

CAPS (N-cyclohexyl-3-aminopropanesulfonic acid) buffer salt is used to formulate CAPS buffer, a zwitterionic buffer that is useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electrotransfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI > 8.5. CAPS is not one of the original Good’s buffers, although it has a similar structure to the other propanesulfonic acids and was selected as a highly water soluble buffering reagent with an optimum buffering pH of 10.4 and minimal reactivity with enzymes or proteins, minimal salt effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CAPS (N-cyclohexyl-3-aminopropanesulfonic acid) buffer salt is used to formulate CAPS buffer, a zwitterionic buffer that is useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electrotransfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI>8.5. CAPS is not one of the original Good’s buffers, although it has a similar structure to the other propanesulfonic acids and was selected as a highly water soluble buffering reagent with an optimum buffering pH of 10.4 and minimal reactivity with enzymes or proteins, minimal salt effects.

Solubility
Soluble in water (100 g/L) at 20°C, and methanol (1%).

Notes
Store at room temperature. Ensure good ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only
Marion J. Sheltawy.; M.S. Losowsky. Determination of faecal bile acids by an enzymic method. Clin. Chim. Acta. 1975, 64 (2),127-132 .