MES, 0.5M buffer soln., pH 5.5
MES, 0.5M buffer soln., pH 5.5
MES, 0.5M buffer soln., pH 5.5
MES, 0.5M buffer soln., pH 5.5
Thermo Scientific Chemicals

MES, 0.5M buffer soln., pH 5.5

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol
Quantity:
100 mL
250 mL
Catalog number J62534.AK
also known as J62534-AK
Price (JPY)
-
Request A Quote
Quantity:
250 mL
Request bulk or custom format
Specifications
CAS145224-94-8
Chemical Name or MaterialMES buffer soln.
ColorColorless
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
View more
MES, is often used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures. It is also been used in various protein characterization assays. This sterile solution suitable for cell culture and molecular biology applications. It is also used as a buffering agent in biology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES, is often used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures. It is also been used in various protein characterization assays. This sterile solution suitable for cell culture and molecular biology applications. It is also used as a buffering agent in biology and biochemistry.

Solubility
Soluble in water (0.5 M at 20°C).

Notes
Solutions are stable at 2-8°C for months. Sterilize by filtration through 0.2 mm filters. Autoclaving is not recommended for any sulfonic acid buffer. If buffers must be nuclease-free, treat the water first, then add the buffer after autoclaving. When MES solutions are autoclaved, they turn yellow (although pH does not change measurably). The identity of the yellow breakdown product is unknown.
RUO – Research Use Only

General References:

  1. BG Bugbee.; FB Salisbury. An evaluation of MES (2 (N-Morpholino) ethanesulfonic acid) and amberlite IRC-50 as pH buffers for nutrient solution studies. Journal of plant nutrition. 1985, 8, ( 7), 567-583.
  2. MJ Alowitz.; MM Scherer . Kinetics of nitrate, nitrite, and Cr (VI) reduction by iron metal. Environmental Science & Technology. 2002, 36, (3), 299-306.

Frequently asked questions (FAQs)

What is the density of MES, 0.5M buffer soln., pH 5.5 (Cat. No. J62534.AK)?

The density of MES, 0.5 M buffer soln., pH 5.5 is 1.07 g/cm3.