Thermo Scientific Chemicals

Cimetidine, 98+%, Thermo Scientific Chemicals

Catalog number: J62825.14
25 g, Each
Thermo Scientific Chemicals

Cimetidine, 98+%, Thermo Scientific Chemicals

Catalog number: J62825.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62825.14
also known as J62825-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
51481-61-9
IUPAC Name
N-cyano-N''-methyl-N'-(2-{[(5-methyl-1H-imidazol-4-yl)methyl]sulfanyl}ethyl)guanidine
Molecular Formula
C10H16N6S
InChI Key
AQIXAKUUQRKLND-UHFFFAOYSA-N
SMILES
CN=C(NCCSCC1=C(C)NC=N1)NC#N
Appearance (Color)
White to off-white
Mass Spec
Conforms
Purity
≥98.0%
Form
Powder

Description

Widely used H2 histamine antagonist which has more recently been described as an inverse agonist. Also a potent I1 imidazoline binding site ligand. It displays antitumor and immunomodulatory activity. Cimetidine competitively inhibits histamine binding to histamine H2 receptors, and suppresses the growth of several tumors, including gastrointestinal cancer. This compound is an anti-angiogenic agent and is an activator of Nischarin. Competitive histamine H2-receptor antagonist which inhibits gastric acid secretion and reduces pepsin output

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble to 50 mM in water and to 100 mM in DMSO, solubility in HCl (almost transparency).

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text