Flunarizine dihydrochloride, 99%
Flunarizine dihydrochloride, 99%
Flunarizine dihydrochloride, 99%
Thermo Scientific Chemicals

Flunarizine dihydrochloride, 99%

CAS: 30484-77-6 | C26H28Cl2F2N2 | 477.42 g/mol
Catalog NumberQuantity
J62969.03
also known as J62969-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62969.03
also known as J62969-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein4284243
CAS30484-77-6
Chemical Name or MaterialFlunarizine dihydrochloride
ColorWhite
EINECS Number250-216-6
View more
A dual sodium/calcium channel blocker; a cerebral and peripheral vasodilator

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only