Lidocaine hydrochloride monohydrate, 98%
Lidocaine hydrochloride monohydrate, 98%
Lidocaine hydrochloride monohydrate, 98%
Thermo Scientific™

Lidocaine hydrochloride monohydrate, 98%

CAS: 6108-05-0 | C14H25ClN2O2 | 288.82 g/mol
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63035.06
also known as J63035-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6108-05-0
IUPAC Name{[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride
Molecular FormulaC14H25ClN2O2
InChI KeyYECIFGHRMFEPJK-UHFFFAOYSA-N
SMILESO.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)White
Thin Layer Chromatography97.5% min.
It is also used as a fast voltage-gated sodium channel blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is also used as a fast voltage-gated sodium channel blocker.

Solubility
Soluble in water (50 mg/ml), chloroform, ethanol, and benzene. Insoluble in diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Do not store above 40 degree C (104 0 F). (preferably store at temperature between 15degree C and 30 degree C).
RUO – Research Use Only

General References:

  1. Thomson, P.D., et al. Lidocaine pharmacokinetics in advanced heart failure, liver disease, and renal failure in humans. Ann. Intern. Med. 1973, 78, (4), 499-508.
  2. Hakim, A.J., et al. Local anaesthetic failure in joint hypermobility syndrome. J R Soc Med. 2005, 98, (2), 84-5.