trans-Zeatin-riboside
trans-Zeatin-riboside
trans-Zeatin-riboside
Thermo Scientific Chemicals

trans-Zeatin-riboside

CAS: 6025-53-2 | C15H21N5O5 | 351.363 g/mol
Quantity:
25 mg
50 mg
100 mg
Catalog number J63382.MC
also known as J63382-MC
Price (JPY)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS6025-53-2
IUPAC Name(2R,3R,4S,5R)-2-(6-{[(2E)-4-hydroxy-3-methylbut-2-en-1-yl]amino}-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular FormulaC15H21N5O5
InChI KeyGOSWTRUMMSCNCW-HNNGNKQASA-N
SMILESCC(CO)=C/CNC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥95.0%
Loss on Drying≤0.5%
Melting Point175.0-180.0°C
View more
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.

Solubility
Soluble in water, ethanol, and methanol.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Carl E. Hansen.; Herman Wenzler.; Frederick Meins Jr.. Concentration Gradients of trans-Zeatin Riboside and trans-Zeatin in the Maize Stem.Plant Physiology.1984, 75, (4), 959-963.
  2. Armstrong, D.J.; Burrows, W.J.; Evans, P.K.; Skoog, F. Isolation of cytokinins from tRNA.Biochemical and Biophysical Research Communications.1969, 37, (3), 451-456.