Methoxychlor
Methoxychlor
Methoxychlor
Thermo Scientific Chemicals

Methoxychlor

Molecule with estrogenic activity | CAS: 72-43-5 | C16H15Cl3O2 | 345.644 g/mol
Quantity:
5 g
25 g
Catalog number J63839.06
also known as J63839-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS72-43-5
IUPAC Name1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
Molecular FormulaC16H15Cl3O2
InChI KeyIAKOZHOLGAGEJT-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(C1=CC=C(OC)C=C1)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Transition Temperature84.0-90.0°C
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥94.0%
Molecule with estrogenic activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only