Cefoperazone sodium salt
Cefoperazone sodium salt
Cefoperazone sodium salt
Thermo Scientific Chemicals

Cefoperazone sodium salt

Cefoperazone sodium salt, CAS # 62893-20-3, is the sodium salt form of cefoperazone. It is a semi-synthetic, broad-spectrum, and third-generation cephalosporin compound with antibiotic and bactericidal activities. | CAS: 62893-20-3 | C25H29N9NaO8S2 | 670.672 g/mol
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65185.03
also known as J65185-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS62893-20-3
IUPAC Namesodium (6R,7R)-7-[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetamido]-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular FormulaC25H26N9NaO8S2
InChI KeyNCFTXMQPRQZFMZ-WERGMSTESA-M
SMILES[Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SCC(CSC4=NN=NN4C)=C(N3C2=O)C([O-])=O)C2=CC=C(O)C=C2)C(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Potency: 870 to 1015 µg/mg (anhydrous basis)
CommentCrystallinity: Conforms to standard
Water Content (Karl Fischer Titration)≤ 5.0%
pH4.5 to 6.5 (0.1% solution in room temperature)
Appearance (Color)White to pale cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• Cefoperazone sodium salt is a semi-synthetic, broad-spectrum, third-generation cephalosporin compound with antibiotic and bactericidal activities
• This compound can inhibit bacterial cell wall synthesis by inactivating penicillin-binding proteins and consequently interfering with the final transpeptidation step required for cross-linking peptidoglycan units, an important component of bacterial cell walls. This leads to a reduction in cell wall stability, causing cell lysis

Application

• Cefoperazone sodium salt exhibits activity against β-lactamase resistant microorganisms
• This compound is also used for studying the expression, binding, and inhibition of penicillin-binding proteins
• Cefoperazone is active in vitro against most gram-positive and gram-negative bacteria, including β-lactamase strains
• This compound has the ability to inhibit ∝-antitrypsin

RUO – Research Use Only

General References:

  1. Neu, H. The in vitro activity, human pharmacology, and clinical effectiveness of new β-lactam antibiotics. Annual Reviews of Pharmacology and Toxicology. 1982, 22, 599-642.
  2. Shahzad, M.; Abbas, A.; Rehman, R.; Mahmud, T.; Shamim, S. Comparative Analysis of Cefotaxime Sodium and Cefoperazone Sodium Drugs Activity and their Phytochemical Combinations Against Bacteria. Asian J. Chem. 2015, 27 (5), 1704-1706.
  3. Hajime, T.; Iwasaki, M.; Ogihara, H. Development of a Novel Chromogenic Medium for Improved Campylobacter Detection from Poultry Samples. J. Food Prot. 2015, 78 (9), 1750-1755.