Caspase-9 Inhibitor III, Thermo Scientific Chemicals
Caspase-9 Inhibitor III, Thermo Scientific Chemicals
Caspase-9 Inhibitor III, Thermo Scientific Chemicals
Caspase-9 Inhibitor III, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Caspase-9 Inhibitor III, Thermo Scientific Chemicals

Catalog NumberQuantity
J65501.MCR
also known as J65501-M^
1 mg
Catalog number J65501.MCR
also known as J65501-M^
Price (JPY)
-
Quantity:
1 mg
Request bulk or custom format
Specifications
CAS403848-57-7
DescriptionWhite
For Use With (Application)A potent and irreversible inhibitor of caspase-9
FormPowder
Formula Weight587.02
View more
A potent and irreversible inhibitor of caspase-9

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C