Midecamycin, 900μg/mg
Midecamycin, 900μg/mg
Midecamycin, 900μg/mg
Thermo Scientific Chemicals

Midecamycin, 900μg/mg

A broad spectrum antibiotic | CAS: 35457-80-8 | C41H67NO15 | 813.979 g/mol
Quantity:
500 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66046.03
also known as J66046-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Boiling Point874°C
CAS35457-80-8
Chemical Name or MaterialMidecamycin
EINECS Number252-578-0
Flash Point>110°C (230°F)
View more
A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only