Piperacillin, 95%
Piperacillin, 95%
Piperacillin, 95%
Thermo Scientific Chemicals

Piperacillin, 95%

CAS: 61477-96-1 | C23H26N5NaO7S | 539.54 g/mol
Quantity:
500 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66143.03
also known as J66143-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS61477-96-1
IUPAC Namesodium (2R,5S,6S)-6-[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular FormulaC23H26N5NaO7S
InChI KeyWCMIIGXFCMNQDS-RAMASXNTSA-M
SMILES[Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@@H]2[C@@H]3SC(C)(C)[C@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Optical Rotation+155.0 to +175.0°
pH2.0-4.5
Assay from Supplier's CofA≥94.0%
Water Content2.0-4.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.

Solubility
Freely soluble in methanol. Only sparingly soluble in aqueous solution at 0.119 mg/mL

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

General References:

  1. Gerald P. Bodey; and Barbara Le Blanc. Piperacillin: In Vitro Evaluation. Antimicrob. Agents Chemother., 1978,14(1), 78-87.
  2. N.A Kuck: G.S. Redin. In vitro and in vivo activity of piperacillin, a new broad-spectrum semisynthetic penicillin. The Journal of Antibiotics, 1978,31(11), 1175-1182.