Dimethyl sulfoxide, Bioreagent
Dimethyl sulfoxide, Bioreagent
Dimethyl sulfoxide, Bioreagent
Dimethyl sulfoxide, Bioreagent
Thermo Scientific Chemicals

Dimethyl sulfoxide, Bioreagent

CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Quantity:
50 mL
100 mL
250 mL
500 mL
1 L
Catalog number J66650.AD
also known as J66650-AD
Price (JPY)
-
Request A Quote
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofATitrable acid: ≤5 mL (≤0.001 meq/g, ≤0.056 mgKOH/g)
Water Content (Karl Fischer Titration)≤0.1%
Appearance (Color)Clear colorless
FormLiquid
Assay from Supplier's CofA≥99.9% (purity by GC)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum.

Solubility
Soluble in water, methanol, acetone, ether, benzene, chloroform.

Notes
Submicron filtered. Store at Room Temperature (15-30°C), desiccate. Hygroscopic.
RUO – Research Use Only