Specifications
Boiling Point582°C
Chemical Name or MaterialGibberellic Acid A4+A7
Flash Point>110°C (230°F)
Recommended StorageStore at -20°C
Solubility InformationSoluble in methanol, ethanol, or acetone; Also soluble in dilute solutions of sodium bicarbonate or sodium acetate
View more