Fibrinopeptide B, Human
Thermo Scientific Chemicals

Fibrinopeptide B, Human

For cellular and molecular biology applications
Quantity:
1 mg
5 mg
Catalog number J66812.MCR
also known as J66812-M^
Price (JPY)
-
Quantity:
1 mg
Request bulk or custom format
Chemical Identifiers
CAS36204-23-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormLyophilized powder
Assay (HPLC)≥98.0%
Assay (special)Peptide content >84%
Solubility1mg/ml in water
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C