D-Ribose-5-phosphate disodium salt hydrate, 85%
D-Ribose-5-phosphate disodium salt hydrate, 85%
D-Ribose-5-phosphate disodium salt hydrate, 85%
Thermo Scientific Chemicals

D-Ribose-5-phosphate disodium salt hydrate, 85%

CAS: 18265-46-8 | C5H9Na2O8P | 274.07 g/mol
Quantity:
250 mg
1 g
Catalog number J66815.MD
also known as J66815-MD
Price (JPY)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS18265-46-8
IUPAC Namedisodium [(2R,3S,4R)-3,4,5-trihydroxyoxolan-2-yl]methyl phosphate
Molecular FormulaC5H9Na2O8P
InChI KeyRHCIGTFSCDIJGH-CHEKPSGWSA-L
SMILES[Na+].[Na+].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow
Assay≥84%
Assay (unspecified)≥84%
Ribose-5-phosphate is an intermediate of the oxidative branch of the Pentose Phosphate Pathway. Ribose-5-phosphate is used in the synthesis of nucleotides and nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ribose-5-phosphate is an intermediate of the oxidative branch of the Pentose Phosphate Pathway. Ribose-5-phosphate is used in the synthesis of nucleotides and nucleic acids.

Solubility
Soluble in water at 50mg/ml

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only