Coelenterazine, native, Aequorea sp., Thermo Scientific Chemicals
Coelenterazine, native, Aequorea sp., Thermo Scientific Chemicals
Coelenterazine, native, Aequorea sp., Thermo Scientific Chemicals
Coelenterazine, native, Aequorea sp., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Coelenterazine, native, Aequorea sp., Thermo Scientific Chemicals

Quantity:
1 mg
10 mg
Catalog number J66823.MA
also known as J66823-MA
Price (JPY)
-
Request A Quote
Quantity:
10 mg
Request bulk or custom format
Specifications
CAS55779-48-1
ChEBICHEBI:2311
Chemical Name or MaterialCoelenterazine
Formula Weight423.46
IUPAC Name8-benzyl-6-(4-hydroxyphenyl)-2-[(4-hydroxyphenyl)methyl]-7H-imidazo[1,2-a]pyrazin-3-one
View more
A sensitive and specific chemiluminescence probe of the superoxide anion

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only