BICINE, Electrophoresis Grade, 99+%
BICINE, Electrophoresis Grade, 99+%
BICINE, Electrophoresis Grade, 99+%
BICINE, Electrophoresis Grade, 99+%
Thermo Scientific Chemicals

BICINE, Electrophoresis Grade, 99+%

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol
Quantity:
25 g
100 g
Catalog number J67087.22
also known as J67087-22
Price (JPY)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Specifications
Beilstein1769362
CAS150-25-4
ChEBICHEBI:40957
Chemical Name or MaterialBICINE
ColorWhite
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only