TA Cloning&trade; Kit, with pCR&trade;2.1 Vector and One Shot&trade; TOP10 Chemically Competent <i>E. coli</i>
Invitrogen™

TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli

The TA Cloning™ Kit with pCR™2.1 vector provides a quick, one-step cloning strategy for directly inserting a Taq-amplified PCR productRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of ReactionsVector
K20404040 ReactionspCR2.1
K20400120 ReactionspCRII
Catalog number K204040
Price (USD)
1,345.65
Online exclusive
1,432.00
Save 86.35
Each
No. of Reactions:
40 Reactions
Vector:
pCR2.1
Price (USD)
1,345.65
Online exclusive
1,432.00
Save 86.35
Each
The TA Cloning™ Kit with pCR™2.1 vector provides a quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into a plasmid vector. The TA Cloning™ Kit uses the pCR™2.1 cloning vector and ExpressLink™ T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

Features of the TA Cloning™ Kit with pCR™2.1 vector:
• Fast & convenient—15-minute, room-temperature ligation
• Efficient—blue/white screening and >80% clones with correct insert
• Flexible—choice of kanamycin or ampicillin resistance for flexible antibiotic selection
• Hassle-free—eliminates any enzymatic modifications of the PCR product
• Streamlined—does not require the use of PCR primers that contain restriction sites

The pCR™2.1 vector provides:
• 3'-T overhangs for direct ligation of Taq-amplified PCR products
• T7 promoter for in vitro RNA transcription and sequencing
• A versatile polylinker with flanking EcoR I sites for easy excision of inserts
• M13 forward and reverse primer sites for sequencing

How TA Cloning™ Works
Taq polymerase has a non-template-dependent activity that adds a single deoxyadenosine (A) to the 3' ends of PCR products. The linearized vector supplied in this kit has single 3' deoxythymidine (T) residues. This allows PCR inserts to ligate efficiently with the vector.

Kit Configurations
The TA Cloning™ Kit is offered in a variety of configurations: without competent cells (K2020-20 and K2020-40), with One Shot™ INVF' Chemically Competent E. coli (K2000-01 and K2000-40), with One Shot™ TOP10F' Chemically Competent E. coli (K2030-01 and K2030-40), and with One Shot™ TOP10 Chemically Competent E. coli (K2040-01 and K2040-40) in 20- and 40-reaction kit sizes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial or Yeast StrainTOP10
Cell TypeChemically Competent E. coli
Cloning MethodTA Cloning
For Use With (Application)PCR Cloning
No. of Reactions40 Reactions
Product LineOne Shot
Product TypeCloning Kit
PromoterT7
Quantity40 reactions
VectorpCR2.1
FormatKit
Unit SizeEach
Contents & Storage
TA Cloning™ kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. The competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.

Citations & References (5)

Citations & References
Abstract
Differential gene expression between developing queens and workers in the honey bee, Apis mellifera [see comments]
Authors:Evans JD, Wheeler DE
Journal:Proc Natl Acad Sci U S A
PubMed ID:10318926
'Many insects show polyphenisms, or alternative morphologies, which are based on differential gene expression rather than genetic polymorphism. Queens and workers are alternative forms of the adult female honey bee and represent one of the best known examples of insect polyphenism. Hormonal regulation of caste determination in honey bees has ... More
An imprinted, mammalian bicistronic transcript encodes two independent proteins.
Authors:Gray TA, Saitoh S, Nicholls RD
Journal:Proc Natl Acad Sci U S A
PubMed ID:10318933
Polycistronic transcripts are common in prokaryotes but rare in eukaryotes. Phylogenetic analysis of the SNRPN (SmN) mRNA in five eutherian mammals reveals a second highly conserved coding sequence, termed SNURF (SNRPN upstream reading frame). The vast majority of nucleotide substitutions in SNURF occur in the wobble codon position, providing strong ... More
PCR-mediated recombination: a general method applied to construct chimeric infectious molecular clones of plasma-derived HIV-1 RNA.
Authors:Fang G, Weiser B, Visosky A, Moran T, Burger H
Journal:Nature Medicine
PubMed ID:9930876
A PCR-based approach was developed that provides a powerful tool for engineering recombinant molecules without reliance on restriction sites. DNA sequences were first amplified by high-fidelity PCR using Pfu polymerase; they were then used both as 'megaprimers' and templates in subsequent asymmetric long PCR amplifications to form chimeric clones. To ... More
Sequence evolution and copy number of Ty1-copia retrotransposons in diverse plant genomes.
Authors: Navarro-Quezada Aura; Schoen Daniel J;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11752395
Sequence evolution of the reverse transcriptase (RT) gene in retrotransposons belonging to the Ty1-copia class was studied in 11 plant species. Phylogenetic reconstruction of the evolutionary history of RT sequences indicated a strong pattern of purifying selection, manifested as high ratios of third to first plus second codon position substitutions, ... More
Immunogene therapy of tumors with vaccine based on Xenopus homologous vascular endothelial growth factor as a model antigen.
Authors: Wei Y Q; Huang M J; Yang L; Zhao X; Tian L; Lu Y; Shu J M; Lu C J; Niu T; Kang B; Mao Y Q; Liu F; Wen Y J; Lei S; Luo F; Zhou L Q; Peng F; Jiang Y; Liu J Y; Zhou H; Wang Q R; He Q M; Xiao F; Lou Y Y; Xie X J; Li Q; Wu Y; Ding Z Y; Hu B; Hu M; Zhang W;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11553767
Overcoming immune tolerance of the growth factors associated with tumor growth should be a useful approach to cancer therapy by active immunity. We used vascular endothelial growth factor (VEGF) as a model antigen to explore the feasibility of the immunogene tumor therapy with a vaccine based on a single xenogeneic ... More