p-Toluenesulfonic anhydride, 97%
p-Toluenesulfonic anhydride, 97%
p-Toluenesulfonic anhydride, 97%
Thermo Scientific Chemicals

p-Toluenesulfonic anhydride, 97%

CAS: 4124-41-8 | C14H14O5S2 | 326.381 g/mol
Quantity:
5 g
25 g
Catalog number L00160.14
also known as L00160-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4124-41-8
IUPAC Name4-methylbenzenesulfonyl 4-methylbenzene-1-sulfonate
Molecular FormulaC14H14O5S2
InChI KeyPDVFSPNIEOYOQL-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale grey to grey
FormCrystals or powder or crystalline powder and/or chunks
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
ClarityClear (5% w/v solution in Acetone)
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only