4-Methoxybiphenyl, 98+%
4-Methoxybiphenyl, 98+%
4-Methoxybiphenyl, 98+%
4-Methoxybiphenyl, 98+%
Thermo Scientific Chemicals

4-Methoxybiphenyl, 98+%

CAS: 613-37-6 | C13H12O | 184.24 g/mol
Quantity:
5 g
25 g
Catalog number L01260.06
also known as L01260-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS613-37-6
IUPAC Name4-methoxy-1,1'-biphenyl
Molecular FormulaC13H12O
InChI KeyRHDYQUZYHZWTCI-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pink
FormCrystals or powder or crystalline powder or lumps
Assay (GC)≥98.0%
Melting Point (clear melt)85.0-91.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only