2-Ethyltoluene, 98+%
2-Ethyltoluene, 98+%
2-Ethyltoluene, 98+%
Thermo Scientific Chemicals

2-Ethyltoluene, 98+%

CAS: 611-14-3 | C9H12 | 120.195 g/mol
Quantity:
5 g
25 g
Catalog number L02028.06
also known as L02028-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS611-14-3
IUPAC Name1-ethyl-2-methylbenzene
Molecular FormulaC9H12
InChI KeyHYFLWBNQFMXCPA-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5020-1.5060 @ 20?C
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyltoluene, is used to predict novel lead compounds for drug discovery that depends on the quality of the docking scoring function.

Solubility
Soluble in water (74.6 mg/L (25°C)).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Monica Alonso.; Laura Cerdan.; Anna Godayol.; Enriqueta Antico.; Juan M. Sanchez. Headspace needle-trap analysis of priority volatile organic compounds from aqueous samples: Application to the analysis of natural and waste waters. Journal of Chromatography A. 2011, 1218, (45), 8131-8139.
  2. A. Roubaud.; R. Minetti.; L.R. Sochet. Oxidation and combustion of low alkylbenzenes at high pressure: comparative reactivity and auto-ignition. Combustion and Flame. 2000, 121, (3), 535-541.